Order Entry
ContactUsLinkComponent
12706 Glass-filled polypropylene female coupling tube to FPT.
Glass-Filled Polypropylene Tube Connectors, Ace Glass Incorporated
Catalog #: 89077-388
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
12706 Glass-filled polypropylene female coupling tube to FPT.
Glass-Filled Polypropylene Tube Connectors, Ace Glass Incorporated
Catalog #: 89077-388
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Description:
    Female Coupling Tube to FNPT (with 3.2 mm Pipe Thread)
  • Cat. no.:
    89077-388
  • Fits Tubing O.D.:
    6.4 mm (¼")
  • Supplier No.:
    12706-18

Specifications

About this item

These glass-filled polypropylene connectors are designed for use with stainless steel, nylon, polyolefin, aluminum, and all other tubing.

Customers Who Bought This Also Bought