Glass-Filled Polypropylene Tube Connectors, Ace Glass Incorporated
Catalog #: 89077-284
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Elbow Tube to Tube
- Cat. no.:89077-284
- Fits Tubing O.D.:9.5 mm (⅜")
- Supplier No.:12705-15
Specifications
About this item
These glass-filled polypropylene connectors are designed for use with stainless steel, nylon, polyolefin, aluminum, and all other tubing.
Customers Who Bought This Also Bought