Order Entry
ContactUsLinkComponent
9436 TUBE, Gas Dispersion
Gas Dispersion Tube, with 10 mm Disc, Ace Glass Incorporated
Catalog #: 89074-516
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
9436 TUBE, Gas Dispersion
Gas Dispersion Tube, with 10 mm Disc, Ace Glass Incorporated
Catalog #: 89074-516
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Porosity:
    C (25 - 50 µm)
  • Description:
    Gas Dispersion Tube
  • Supplier No.:
    9436-06

Specifications

About this item

Gas dispersion tube has a disc diameter of 10mm, and an overall length of 150mm

The disc is approximately 2mm thick. Outside diameter of the tube is 7mm. Available in five different porosities. Fabricated from borosilicate glass.

Customers Who Bought This Also Bought