Septa, Sleeve Type, Rubber, Ace Glass Incorporated
Catalog #: 89064-974
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Color:Red
- Description:For [ST]19/38, [ST]19/22 Joints
- Cat. no.:89064-974
- Supplier No.:9096-54
Specifications
About this item
This sleeve-type septa has a hollow plug
The top is flanged with a sleeve-like extension that folds down over the neck of vessel. The diaphragm can be punctured readily with a syringe needle. Puncture seals automatically after the needle is withdrawn.
Ordering information: Rubber septum are available in white, gray, or red finishes.
Customers Who Bought This Also Bought