Order Entry
ContactUsLinkComponent
8801 STANDARD WALL TUBING, Borosilicate - Duran
Borosilicate Glass Standard Wall Tubing, Ace Glass Incorporated
Catalog #: 89064-138
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
8801 STANDARD WALL TUBING, Borosilicate - Duran
Borosilicate Glass Standard Wall Tubing, Ace Glass Incorporated
Catalog #: 89064-138
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Tubing O.D.:
    300 mm
  • Wall thickness:
    7 mm
  • Cat. no.:
    89064-138
  • Supplier No.:
    8801-345

Specifications

About this item

Tubing comes in lengths of 1.5 meters.

Customers Who Bought This Also Bought