Order Entry
ContactUsLinkComponent
8645 TUBE, Test, Stoppered
Test Tubes, Ace Glass Incorporated
Catalog #: 89063-144
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Test Tubes, Ace Glass Incorporated
Catalog #: 89063-144
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Dimensions:
    16 x 150 mm (5/8 x 6")
  • Description:
    Stopper (only), #16
  • Cat. no.:
    89063-144
  • Supplier No.:
    8645-04

Specifications

About this item

Test tubes, available with or without matching glass stoppers; [ST] 24/40 stopper is hollow; #16 and #19 stoppers are solid.

Fabricated from borosilicate glass.

Ordering information: Available complete with stopper and tube, or components can be purchased separately.
Customers Who Bought This Also Bought