O-Ring Replacement Sets for Stopcocks, Ace Glass Incorporated
Catalog #: 89062-456
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Kalrez O-Rings
- Stopcock Size:0 - 3, 0 - 4 mm
- Supplier No.:8194-124
Specifications
About this item
Replacement O-Ring sets are designed for use with stopcock plugs.
Available in five different compounds.
Packaging: Packaging includes two large O-Rings and one small O-Ring for the plug sizes specified.
Customers Who Bought This Also Bought