Stopcock Plug Pullers, Ace Glass
Catalog #: 89062-454
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Stopcock Plug Puller
- Cat. no.:89062-454
- For Use With:0 – 15 mm stopcocks
- Supplier No.:8194-115
Specifications
About this item
Stopcock plug pullers are used to remove the PTFE plug from the threaded barrel in old style (see 89062-406 series) threaded stopcocks.
Plug pullers are not compatible with hooded style stopcocks (as supplied in 89062-594 series). The flat nylon disc has a matching thread in the center, a slit on one side, and a spring steel wire in a groove on the outside edge. Simply spread the disc over the thread on the plug, effectively increasing the thread length, and turn the plug until the O-rings release.
Customers Who Bought This Also Bought