O-Rings, FETFE®, Ace Glass Incorporated
Catalog #: 89061-016
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:FETFE O-Rings
- Cat. no.:89061-016
- Supplier No.:7855-734
Specifications
About this item
FETFE is an exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along with excellent temperature and chemical compatibility. Generally black or white in color.
Customers Who Bought This Also Bought