ContactUsLinkComponent
7855 O-RING
O-Rings, Viton, Ace Glass Incorporated
Catalog #: 89060-626
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
7855 O-RING
O-Rings, Viton, Ace Glass Incorporated
Catalog #: 89060-626
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Description:
    Viton O-Rings
  • Cat. no.:
    89060-626
  • Supplier No.:
    7855-16
Specifications
About this item
Viton o-rings
Viton is a class of elastomers comprised of linear co-polymer of vinylidine flouride and hexafluoropropylene. Fluorine content is between 66 to 70%. Generally black in color, viton o-rings provide generally good temperature and chemical resistance.
Customers Who Bought This Also Bought