Rodaviss® O-Rings, Ace Glass Incorporated
Catalog #: 89059-728
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:19/22 Rodaviss® O-Rings, Nitrile
- Cat. no.:89059-728
- Supplier No.:7617-04
Specifications
About this item
Designed for use with Rodaviss® joints and caps to form a leak-tight seal
Available in Nitrile or Viton formats. For threaded joints, see 89059-696 series. For caps, see 89059-718 series.
Customers Who Bought This Also Bought