Bushings, Ace Glass
Catalog #: 89059-246
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Tubing I.D.:47-48 mm
- Description:PTFE Bushing, FETFE O-Ring
- Cat. no.:89059-246
- Includes:O-Ring 89061-030
- Thread Size:#50
- Supplier No.:7506-35
Specifications
About this item
Front-seal bushing connector for use with Ace-Thred joints, threaded glass, or stainless steel connectors
Used to join threaded end to a reduced-end tube. Available in either nylon or PTFE.
Customers Who Bought This Also Bought