Flask, Filtering, Heavy Wall, Ace Glass Incorporated
Catalog #: 89057-382
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Capacity:2000 ml
- Description:Flask (only)
- Cat. no.:89057-382
- Stopper No.:11
- Supplier No.:6983-20
Specifications
About this item
This flask has a tooled neck for a uniform stopper fit
The side hose connection is a #11 Ace-Thred that accepts a PTFE tubing connector with a silicone o-ring and nylon bushing to make an easier, safer, more convenient vacuum hose connection. To use, simply slide the bushing over the connector, attach hose to serrated end, and insert into Ace-Thred and tighten until the o-ring compresses, making a leak-tight seal. Fabricated from borosilicate glass.
Ordering information: Complete item consists of appropriate volume flask, 1/4-inch (89052-940) or 3/8-inch (89052-942), #11 Ace-Thred tubing connector with 89060-646 silicone o-ring size -011, and 89059-212 nylon bushing. Components may also be purchased separately. Rubber stopper is NOT included.
Customers Who Bought This Also Bought