Order Entry
ContactUsLinkComponent
6252 DESICCATOR CABINET, Small - BelArt
Desiccator Cabinet, Horizontal, Clear, Ace Glass
Catalog #: 89054-062
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
6252 DESICCATOR CABINET, Small - BelArt
Desiccator Cabinet, Horizontal, Clear, Ace Glass
Catalog #: 89054-062
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Description:
    Desiccator Cabinet, Horizontal Profile Clear
  • Exterior dimensions:
    34.1W×41D×21.3H cm (13.4×16.3×8.4"). Door: 22.5W×31H cm (8.9×12.3")
  • Int. volume:
    0.75 cu. ft.
  • Cat. no.:
    89054-062
  • Supplier No.:
    6252-25

Specifications

About this item

This stackable, clear plastic Secador™ desiccator cabinet is perfect for cramped locations

Can be safely stacked up to three high. Cabinet is completely transparent with built-in Hygrometer. Patented sealed construction; for air-tight, dust- and moisture-free storage. One latch; provision for lock or tamper evident seal. Use any desiccant.

Customers Who Bought This Also Bought