Ace-Safe Tubing Connector, Polypropylene, Ace Glass Incorporated
Catalog #: 89052-942
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Tubing I.D.:3/8"
- Description:Polpropylene Tubing Connector (only)
- Cat. no.:89052-942
- Thread Size:#11
- Supplier No.:5853-10
Specifications
About this item
These polypropylene tubing connectors are used with nylon bushings to make an easy and safe connect/disconnect of coolant or steam tubing to jacketed reaction flasks, condensers, or any vessel utilizing the Ace-Thred as the connection port
Connector is fitted with a silicone o-ring. Bushing compresses connector and o-ring for leak-tight seal. Once tubing is attached to serrated end of connector, bushing will not come off. Polypropylene connectors have a maximum temperature of 110°C.
Ordering information: Available complete with bushing, connector, and o-ring, or as polypropylene connector only. Also available in glass-filled PTFE. Contact your Avantor representative for more information.
Customers Who Bought This Also Bought