Boston Bottle, Round, Safety Coated, Ace Glass
Catalog #: 89051-852
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Cap Size:38-439C
- Capacity:2.4 L (80 oz.)
- Description:Boston Round Safety Coated Clear Bottle, with Jug Handle
- Cat. no.:89051-852
- Diameter ×H:135 x 291 mm
- Supplier No.:5546-20
Specifications
About this item
Clear glass bottles, fabricated of soda lime, flint glass.
- Not autoclavable
- Supplied without caps
PVC safety coated to reduce potential breakage, and to prevent exposure to laboratory personnel in handling hazardous materials.
Customers Who Bought This Also Bought