ContactUsLinkComponent
5420 BOTTLE, Specific Gravity
Bottle, Specific Gravity, Ace Glass Incorporated
Catalog #: 89051-706
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
5420 BOTTLE, Specific Gravity
Bottle, Specific Gravity, Ace Glass Incorporated
Catalog #: 89051-706
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Capacity:
    25
  • Description:
    Specific Gravity Bottle, Complete
  • Cat. no.:
    89051-706
  • Supplier No.:
    5420-24
Specifications
About this item
This Guy-Lussac type bottle is unadjusted, ready for calibration in the laboratory
When adjusted, it is suitable for ASTM D 369. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought