Hose Connection Adapter, Bent, Ace Glass Incorporated
Catalog #: 89051-402
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Hose Connection Adapter with Standard Taper Joint
- Cat. no.:89051-402
- Hose Connections:3/8" (D)
- Joint Size:[ST] 19/38
- Supplier No.:5205-05
Specifications
About this item
These Hose Connection Adapters are available with either a standard taper inner or a spherical ball bottom joint.
- Inner [ST] or spherical ball bottom joint
- 90° angle top hose connection
- Fabricated from borosilicate glass
At the top of the adapter is a bent 90° hose connection. Fabricated from borosilicate glass. Also available with an integral Porosity A (145–174 micron) fritted disc.
Customers Who Bought This Also Bought