Specifications
- Description:pH-Fix pH Test Strips, 6×85 mm
- Cat. no.:89049-924
- pH Range:0.0 - 14.0
- pH Graduation:1.0
- Supplier No.:92110
- No. of Strips:100
Specifications
About this item
pH-Fix test strips allow fast pH testing directly at the point of interest.
- Ready to use
- Easy and safe
- Packed in classic flat box
- Several test pads for exact results
- Safe analysis by long plastic handle
- Fixed indicator dye does not leach into sample
Even in weakly buffered solutions, a safe and reliable pH determination becomes possible, as the strips can be immersed for a long time until the final reaction color has developed
pH-Fix test strips are tested and approved for medical applications and carry a CE-mark for in-vitro-diagnostics 98/79/EG or medical products 93/42/EWG. They meet the special demands of health care professionals and ensure safe results for medical pH-testing.
The colors of the individual test pads don't bleed and mix, which assures exact comparison to the color scale.