Order Entry
ContactUsLinkComponent
VWR Unifrost Slide
Microscope Slides, Frosted Glass, Azer Scientific
Catalog #: 89049-670
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
VWR Unifrost Slide
Microscope Slides, Frosted Glass, Azer Scientific
Catalog #: 89049-670
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise

Specifications

  • Color:
    White
  • Description:
    Sysmex slides
  • L×W:
    75×25 mm (3×1")
  • Thickness:
    1.0 mm

Specifications

About this item

These frosted microscope slides are specifically designed for compatibility with Sysmex® systems

  • Clipped corners at 45°
  • Single-frosted edges
  • Sysmex® systems compatible
  • High-quality glass

VWR frosted microscope slides are specifically designed for compatibility with Sysmex® systems, they are manufactured from high-quality glass. These slides are pre-cleaned with smooth grounded edges and undergo continuous premium quality processing. All slides have single side, color frosted ends which helps to avoid sticking and jamming problems that can occur with other slides. The color frosted end is resistant to all common chemicals that are utilized in the laboratory. Slides feature 45° clipped corners for ease in handling, and two choices of color-coded frosted ends to aid in quick and simple identification.

They are manufactured from high-quality glass. Slides are pre-cleaned with smooth grounded edges and undergo continuous premium quality processing. All slides have single side, color frosted ends which help to avoid sticking and jamming problems that can occur with other slides. The color frosted end is resistant to all common chemicals that are utilized in the laboratory. Slides feature 45° clipped corners for ease in handling, and two choices of color-coded frosted ends to aid in quick and simple identification.

Packaging: 72 per box, 20 boxes per case
Customers Who Bought This Also Bought