SET OF CALIBRATION SUBSTANCES
Catalog #: 89034-736
Supplier: Buchi
Customers Who Bought This Also Bought
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$214.30 to $2,349.04
Water ACS, Reagent Grade ASTM Type I, ASTM Type II Packaged in plastic containers
Supplier: Ricca Chemical
$200.03
Acetic acid glacial, anhydrous 100% (by alkalimetry), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
$394.61 to $1,493.28