Order Entry
ContactUsLinkComponent
VWR Stirring Bar, 25x14mm
VWR® Magnetic Rare Earth Stir Bars
Catalog #: 89030-554
Supplier:  VWR International
VWR Stirring Bar, 25x14mm
VWR® Magnetic Rare Earth Stir Bars
Catalog #: 89030-554
Supplier:  VWR International

Specifications

  • Coating:
    PTFE
  • Ø:
    14 mm (9/16")
  • Length:
    25 mm (1")
  • Cat. no.:
    89030-554
  • Shape:
    Elliptical

Specifications

About this item

Molded in pure, FDA-approved PTFE, these samarium-cobalt elliptical magnets feature increased magnetic strength and are almost totally resistant to demagnetization.

  • Autoclavable
  • PTFE construction
  • Highly resistant to demagnetization

Rare Earth magnets are identified by a chemically inert carbon black spot.

Customers Who Bought This Also Bought