Order Entry
ContactUsLinkComponent
PTFE Plug
Accessories for Eight-Neck Reaction Vessel Lids and Adapters, 200 mm, Chemglass
Catalog #: 89030-000
Supplier:  Chemglass
Small Business Enterprise
PTFE Plug
Accessories for Eight-Neck Reaction Vessel Lids and Adapters, 200 mm, Chemglass
Catalog #: 89030-000
Supplier:  Chemglass
Small Business Enterprise

Specifications

  • Description:
    Replacement PTFE plug, 1.9 cm (³/₄") male NPT
  • Cat. no.:
    89030-000
  • Supplier No.:
    CG-1945-T-92

Specifications

About this item

Customers Who Bought This Also Bought