Order Entry
ContactUsLinkComponent
Drum & Carboy Funnel
Drum and Carboy Funnels, Chemglass
Catalog #: 89028-778
Supplier:  Chemglass
Small Business Enterprise
Drum & Carboy Funnel
Drum and Carboy Funnels, Chemglass
Catalog #: 89028-778
Supplier:  Chemglass
Small Business Enterprise

Specifications

  • Capacity:
    4.3 L
  • Cat. no.:
    89028-778
  • Top Diameter:
    260 mm
  • Supplier No.:
    CG-1766-02

Specifications

About this item

Large funnels with handles are ideal for filling columns, barrels, tanks, and carboys providing ample volume and fast flow

The two smaller sizes have a handle at the rim. Molded of natural polypropylene, funnels are autoclavable.

Customers Who Bought This Also Bought