Bottle Cap Assembly, Chemglass
Catalog #: 89028-400
Supplier: Chemglass
Small Business Enterprise
Specifications
- Color:Black
- Description:Cap Assembly for GPI Threaded Bottles, PP
- Cat. no.:89028-400
- Thread Size:1/8"
- Supplier No.:CG-1158-15
Specifications
About this item
Bottle cap for GL-45 and GPI-38 threads that permits standard solvent or media bottles to be used as an HPLC reservoir.
- Caps are made from polypropylene
- Inserts are made from tefzel
Complete cap assemblies consists of the cap, tefzel insert, two plugs for the 1/8" tubing openings and a plug for the opening. Caps have two holes to accomodate 1/8" tubing and one hole that accepts a standard luer inner joint.
Customers Who Bought This Also Bought