EP Scientific Wide Mouth Packers, Amber Glass, Thermo Scientific
Catalog #: 89026-606
Supplier: Thermo Scientific
Specifications
- Cap Size:70-400
- Capacity:2.5 L (80 oz.)
- Color:Amber
- Description:Bottle, amber glass, wide mouth packer with cap, depyrogenated
- Cat. no.:89026-606
- O.D.×H:14 x 23.9 cm (3.9 x 7.3'')
- Supplier No.:12380APF
Specifications
About this item
These wide mouth packers are preferred for solids, semi-solids and hazardous waste samples.
- Amber bottles protect light-sensitive materials
- Class 100/10 glass
- Certificate of Analysis is included with each shipment
- Recommended for endotoxin-free applications
Particle-certified glass containers are tested and certified to meet the low particle criteria; assembled to contain as few as five particles per milliliter >0.5 µm.
Depyrogenated glass containers are specially prepared to meet endotoxin levels of less than 0.06 EU/ml for use in terminal sterilization, water sampling and product storage.
Recommended for low particle applications. Individual bottles double bagged to maintain integrity until point of use.
Ordering information: Custom cleaning services for additional applications are available; contact your Avantor sales representative for more information.
Delivery: All bottles are supplied with attached caps. Choose from solid-top polypropylene caps with fluoropolymer resin liners or open-top polypropylene caps with fluoropolymer resin-backed silicone septum ultrasonically welded into place.
Customers Who Bought This Also Bought