Order Entry
ContactUsLinkComponent
VWR* PTFE Joint Clamps
VWR® Glass Joint Clamps, PTFE
Catalog #: 89026-180
Supplier:  VWR International
VWR* PTFE Joint Clamps
VWR® Glass Joint Clamps, PTFE
Catalog #: 89026-180
Supplier:  VWR International

Specifications

  • Cat. no.:
    89026-180
  • ST Joint Size:
    24/-

Specifications

About this item

PTFE joint clamps can withstand temperatures of up to 260°C (500°F).

  • Chemically resistant and inert

Feature PTFE-encapsulated steel springs that can be adjusted by hand.

Customers Who Bought This Also Bought