Specifications
- Length:3 1/2"
- Description:Surgi OR™ Backhaus Towel Clamps
- Material:Stainless Steel
- Tip Configuration:Sharp
- Curvature:Curved
- Cat. no.:89023-684
- Supplier No.:95-606
Specifications
About this item
Surgi-OR™ Backhaus Towel Clamps are a perforating clamp used for grasping tissue, securing towels or drapes and holding or reducing small bone fractures
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR™ instruments are a cost-effective line of instruments that must meet Sklar's quality standards.