To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Length:27.9 cm (11")
- Width:20.3 cm (8")
- Description:System
- Cat. no.:89012-492
- Supplier No.:901806
Specifications
About this item
Compact, portable tabletop system can safely anesthetize animals up to 13.6 kg (30 lbs.). The small footprint makes it ideal for areas with limited floor and counter space
- Single dial setting delivers a consistent and safe anesthetic level
- Breathing circuits allow flat, prone, or supine positions with head access
- All waste gases are scavenged for easy evacuation. Positive sealing gaskets on induction chambers minimize exposure to any waste gases
- Main unit has a solid aluminum frame with a baked enamel finish that resists scratching and staining
The system can be connected to in-house evacuation, fume hood, or disposable charcoal filter.