VWR® Glove Box Holders, Acrylic
Catalog #: 89011-724
Supplier: VWR International
Specifications
- Capacity:3 Boxes
- Cat. no.:89011-724
- W×H×D:38.5x29.5x11 cm (153/16x115/8x45/16")
Specifications
About this item
Dispensers are constructed of tough, clear acrylic to provide visibility of glove size and material.
Dispensers are constructed of tough, clear acrylic to provide visibility of glove size and material. Each holder includes a polyester foam insert to securely hold a range of glove boxes while ensuring the easy removal of gloves. Supplied with wall-mounting screws.
- Constructed of tough, clear acrylic
- Polyester foam insert securely holds glove boxes
Each holder includes a polyester foam insert to securely hold a range of glove boxes while ensuring the easy removal of gloves. Supplied with wall-mounting screws.
Customers Who Bought This Also Bought