SP Bel-Art Secador® Auto-Desiccator Module Desiccant Replacement Beads, Bel-Art Products, a part of SP
Catalog #: 89009-210
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:26 g Bottle to fill Secador® Auto-Desiccator Modules
- Cat. no.:89009-210
- Supplier No.:420740020
Specifications
About this item
Color-indicating silica gel beads for use in refilling the Secador® auto-desiccator module.
- Color-changing moisture indicator beads
- Inert, non-toxic, and non-corrosive
- Replacement beads indicate moisture saturation and replacement time
Silica gel absorbs moisture from the air and is inert, non-toxic and non-corrosive
Beads contain a color-changing moisture indicator that turns from blue to light pink, indicating that the silica gel is saturated and can not absorb additional moisture.
Secador® auto-desiccator modules automatically regenerate the silica gel beads for thousands of cycles; however, when beads turn pink/dark brown, and fail to regain their blue color, it is time to replace the beads with a fresh supply.
Unlike other desiccants, silica gel is non-friable and won’t disintegrate into powder even after repeated regeneration.
Customers Who Bought This Also Bought