STAT Biohazard Bag with Document Pouch
Catalog #: 89005-358
Supplier: Minigrip
Specifications
- Dimensions:15.2×22.8 cm (6×9")
- Thickness:2 mil
- Cat. no.:89005-358
- Supplier No.:MGTZ69S
Specifications
About this item
Easily identifiable bag is constructed of red film and features the term STAT printed on the front.
- TearZone® access for simple, safe use.
Bag front also features a biohazard logo and check boxes labeled Freeze, Refrigerate, and Room Temperature.
Delivery: Supplied in easy access perforated dispenser bags and can be easily ripped open for efficiency.
Customers Who Bought This Also Bought