Order Entry
ContactUsLinkComponent
TUBE DCT NOCAP 16X150MM CS1000
KIMAX® Culture Tubes, Disposable, with Screw Thread Finish, Kimble Chase
Catalog #: 89003-572
TUBE DCT NOCAP 16X150MM CS1000
KIMAX® Culture Tubes, Disposable, with Screw Thread Finish, Kimble Chase
Catalog #: 89003-572

Specifications

  • Capacity:
    20 ml
  • Ø:
    16 mm
  • Height:
    150 mm
  • GPI Thread Finish:
    15–415
  • Supplier No.:
    73750-16150

Specifications

About this item

Tubes are suitable for tissue culture work and general bacteriological use.

Tubes are suitable for tissue culture work and general bacteriological use.

Tubes are made from KIMAX-51® borosilicate glass in compliance with ASTM E438, Type I, Class A or B glass requirements. Each tube has a chevron marking area. Tubes are shrink-wrapped in plastic film in modular trays. Supplied without caps.

Customers Who Bought This Also Bought