Phenolic Screw Caps with Rubber Liner, Kimble Chase
Catalog #: 89003-552
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Cap Size:13-415
- Cat. no.:89003-552
Specifications
About this item
Phenolic cap material and cemented-in rubber liner are resistant to the effects of repeated autoclaving
Suitable for use with 89003-568 series disposable culture tubes and 21020-607 series disposable centrifuge tubes. Designed from ASTM Specification E982.
Customers Who Bought This Also Bought