Phenolic Screw Thread Closure, Kimble Chase
Catalog #: 89001-506
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Cap Size:24-410
- Color:Black
- Height:19 mm
- Cat. no.:89001-506
- Supplier No.:45066C-24
Specifications
About this item
Caps have mineral-filled phenolic material to withstand the effects of repeated autoclaving.
Rubber-lined caps retain adhesive properties and meet ASTM Specification E982, Class A requirements. PTFE-faced rubber-lined caps meet ASTM specification E982, Class B requirements.
Unlined closures have excellent chemical resistance and are autoclavable.
Customers Who Bought This Also Bought