Phenolic Screw Thread Closure, Kimble Chase
Catalog #: 89001-048
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Cap Size:15-415
- Color:Black
- Height:16 mm
- Cat. no.:89001-048
- Supplier No.:45066B-15
Specifications
About this item
Caps have mineral-filled phenolic material to withstand the effects of repeated autoclaving.
Rubber-lined caps retain adhesive properties and meet ASTM Specification E982, Class A requirements. PTFE-faced rubber-lined caps meet ASTM specification E982, Class B requirements.
Unlined closures have excellent chemical resistance and are autoclavable.
Customers Who Bought This Also Bought