Polypropylene Screw Caps, BrandTech
Catalog #: 83009-034
Supplier: Brandtech
Specifications
- Cap Size:GL32
- Cat. no.:83009-034
- Supplier No.:V83312
Specifications
About this item
Translucent screw caps are for use with BrandTech polypropylene and low-density polyethylene bottles.
- Autoclavable translucent polypropylene
- Fits VITLAB and many other bottles with standard thread
Packaging: Bulk packed.
Customers Who Bought This Also Bought