ThinCert™ Tissue Culture Inserts, Sterile, Greiner Bio-One
Catalog #: 82051-568
Supplier: Greiner Bio-One
Product available on GSA Advantage®
Specifications
- Pore size:1.0 µm
- Density:2 pore×10⁶ cm²
- Optical Properties:Transparent
- Supplier No.:657610
Specifications
About this item
Tissue culture inserts provide optimal support for co-cultivation, cell migration, and transport process applications.
- Sterile
- Spacers eliminate capillary effect
- Individually wrapped inserts
- Low protein binding capacity
- Improved cell adhesion
Advanced design permeable inserts feature transparent or translucent polyethylene terephthalate (PET) membranes that offer low protein binding capacity, improved cell adhesion, and defined pore size to ensure complete exchange of compounds and nutrients
ThinCert™ membranes are sealed to a robust, transparent polystyrene housing. Housing features self-positioning hanging geometry with eccentric in-well positioning, improving both the pipetting angle and window. Spacers maintain distance to eliminate capillary effect. Inserts are individually wrapped.
Membrane will not curl after detachment.
Customers Who Bought This Also Bought