384-Well No-Bottom Plates, Greiner Bio-One
Catalog #: 82051-544
Supplier: Greiner Bio-One
Specifications
- Color:Black
- Cat. no.:82051-544
- Supplier No.:781000-06
Specifications
About this item
Standard and Small Volume™ LoBase plates are made from virgin polystyrene resin and molded without the bottom of the plate.
- Smaller working volume reduces sample usage
- Chimney-well design helps prevent cross-contamination
- Virgin polystyrene construction
- Molded without a bottom
The Small Volume format features far smaller working volumes than typical 384-well plates, reducing sample usage while avoiding the instrument compatibility issues introduced by 1536-well plates. The LoBase format features a chimney-like arrangement of wells in which each well stands on its own to reduce the possibility of cross-contamination. Chimney wells also feature individual rims that rise above the surface of the plate to aid in sealing by cap mat, thermal sealer, or adhesive sealer.
Customers Who Bought This Also Bought