Media Bottles, PET, Sterile, Greiner Bio-One
Catalog #: 82051-494
Supplier: Greiner Bio-One
Product available on GSA Advantage®
Specifications
- Capacity:100 ml (3.4 oz.)
- Cat. no.:82051-494
- Brim Capacity:125 ml
- Supplier No.:951700
Specifications
About this item
These sterile PET media bottles are ideal for storage of media, buffers, and laboratory samples.
- Clear polyethylene terephthalate construction
- Sturdy and crack-resistant
- Flexible
- Free of RNase, DNase, and human DNA
- Nonpyrogenic and noncytotoxic
The square design of the media bottles allows efficient use of storage space.
Clear polyethylene terephthalate (PET) construction makes content identification simple. The sturdy yet flexible crack-resistant bottles are supplied with white screw caps. All bottles are graduated to the working volume and supplied sterile.
Customers Who Bought This Also Bought