ContactUsLinkComponent
384-Well Storage Plate
MASTERBLOCK® 384-Well Deep Well Microplates, Greiner Bio-One
Catalog #: 82051-320
Supplier:  Greiner Bio-One
Product available on GSA Advantage®
384-Well Storage Plate
MASTERBLOCK® 384-Well Deep Well Microplates, Greiner Bio-One
Catalog #: 82051-320
Supplier:  Greiner Bio-One
Product available on GSA Advantage®
Specifications
  • Sterility:
    Non sterile
  • Color:
    Clear
  • Cat. no.:
    82051-320
  • Working Volume:
    20 - 225 µl
  • Supplier No.:
    781270
  • Total Volume:
    240 µl
Specifications
About this item
Microplates are manufactured from ClearChoice™ virgin polypropylene resin and produced with uniform external dimensions and tolerances to meet ANSI recommendations for compatibility with equipment and robotics.
  • Virgin polypropylene construction
  • Internal well geometry to minimize wicking and air bubble formation
  • Autoclavable
  • Supplied without lids
  • High temperature tolerance
  • Biologically inert
These polypropylene microplates are biologically inert, exhibit low binding characteristics, offer a high temperature tolerance, and are resistant to many standard laboratory chemicals
Microplates feature square-round internal well geometry to minimize wicking and air bubble formation. Conical well bottoms facilitate complete sample recovery, and the unique design and well depth enable many popular styles of pipet tips complete access to the plate bottom. Raised well rims aid in thermal, adhesive, or cap mat sealing. All plates are autoclavable and supplied without lids.
Microplates are RNase-free, DNase-free, pyrogen-free, and free of DNA-polymerase inhibitors.
These properties provide optimal conditions for laboratory specimen and compound storage.
Customers Who Bought This Also Bought