Inoculating Loops and Needles, Sterile, Greiner Bio-One
Catalog #: 82051-142
Supplier: Greiner Bio-One
Product available on GSA Advantage®
Specifications
- Length:20 cm (77/8")
- Color:Blue
- Description:10 µL Inoculating Loops
- Cat. no.:82051-142
- Packaging:50/Bag
- Supplier No.:731170
Specifications
About this item
Inoculating loops allow easier bacterial collection and inoculation.
- Color-coded
- Polystyrene construction
- Sterile
Inoculating needles are ideal for picking up single colonies.
Customers Who Bought This Also Bought