CELLCOAT® Cell Culture Microplates, Greiner Bio-One
Catalog #: 82050-850
Supplier: Greiner Bio-One
Specifications
- Well Volume:16.1 mL
- Description:6-Well
- Working Well Volume:2 - 5 ml
- Plate Color:Clear
- Bottom Color:Clear
- Supplier No.:657940
Specifications
About this item
Convenient, ready-to-use polystyrene cell culture plates are coated with extracellular matrix (ECM) proteins or synthetic polypeptides.
- Polystyrene construction
- Compact footprint
- Supplied with lid
- Convenient and ready to use
These molecules provide an inert scaffolding that participates directly and indirectly in the regulation of cell growth and proliferation. Ideal for serum-free and serum-reduced cultivation or for differentiation of individual cell types. These multiwell tissue culture plates are consistently coated with Collagen Type 1 (from Rat Tail Tendon), Poly-D-Lysine or Poly-L-Lysine (both synthetic), and feature a “Best Before” date and lot number. All plates are supplied with lids and have a footprint conforming to the ANSI/SBS 1-2004 Standard.
Dimensions: 128L x 86W mm
Customers Who Bought This Also Bought