Specifications
- Sterility:Non sterile
- Color:Clear
- Description:Standard Lids with Condensation Rings
- Material:Polystyrene
- Supplier No.:656170
Specifications
About this item
Made of polycarbonate or polystyrene resin, these lids are best suited for chimney-well style 96-well plates in cell culture applications.
- Polycarbonate or polystyrene resin construction
- Fully skirted lids
- Free of RNase and DNase
- Nonpyrogenic
Half-area plates maintain ANSI/SBS standards and well-to-well pitch, yet feature a well diameter design that offers one-half the well surface area of standard plates
Standard full skirt lids allow easy gripping of the lid manually or robotically. Low-profile lids feature a partial skirt, facilitating robotic gripping of the plate while the lid remains on. Two corner notches on each lid assure proper plate-to-lid orientation, minimizing cross-contamination. Lids with condensation rings minimize both evaporation and cross-contamination. Black opaque lids are ideal when a loose cover is required to shield light-sensitive assays or compounds.
Lids are RNase-free, DNase-free, and nonpyrogenic.
This format allows a 50% sample volume reduction without moving to high well density plates. Half-area plates feature a medium binding polystyrene surface, which is ideal for work with nonpolar proteins and peptides.