96-Well Microplates, Polystyrene, Clear, Greiner Bio-One
Catalog #: 82050-720
Supplier: Greiner Bio-One
Product available on GSA Advantage®
Specifications
- Color:Clear
- Cat. no.:82050-720
- Well Shape:Flat-Bottom
- Surface Treatment:High binding
- Supplier No.:655061
Specifications
About this item
These 96-well microplates are available with flat-, round-, conical-, and C-bottom well styles, and are manufactured from clear, virgin polystyrene and selected for optical clarity and consistency.
- Clear virgin polystyrene construction
- High degree of optical clarity
- Chimney-well design helps reduce cross-contamination
- Designed to minimize air bubble formation
C-bottom plates feature a flat bottom for optical measurement, but the hard corners are curved to eliminate liquid traps and improve plate washing
All plates are free of detectable RNase, DNase, human DNA, and pyrogens.
MICROLON™ 200 and MICROLON™ 600 formats are available for consistent performance in binding assays. The chimney-well design option reduces the risk of cross-contamination and facilitates sealing by cap mat, thermal sealer, or adhesive sealer. Plates are supplied without lids.
Customers Who Bought This Also Bought