To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- L×W:75×25 mm
- Thickness:1 mm
- Cat. no.:82030-480
- Chamber Volume:10 µl
- Chamber Depth:100 µm
- Grid Type:Burker Turk
- Supplier No.:DHCB025
Specifications
About this item
These plastic hemacytometers offer repeatable and reliable measurements and analysis
The C-Chip™ hemacytometers are biocompatible and optically transparent. The light and sturdy construction is resistant to surface scratches or cracks. The hemacytometer consists of two surface-patterned, enclosed chambers with two ports for sample introduction. Results are readable under microscopic observation after one to three minutes have elapsed.
These plastic hemacytometers offer repeatable and reliable measurements and analysis
The C-Chip™ hemacytometers are biocompatible and optically transparent. The light and sturdy construction is resistant to surface scratches or cracks. The hemacytometer consists of two surface-patterned, enclosed chambers with two ports for sample introduction. Results are readable under microscopic observation after one to three minutes have elapsed.
The unique design eliminates the use of coverslips and allows for exact volume control. This reduces the user’s risk of exposure to infectious or hazardous samples, and increases efficiency throughout laboratory procedures by eliminating the need for washing or autoclaving components. Available in four different grid types for a broad range of applications, including blood cell counting, cell concentration measurement, cell viability testing, bacteria and fungal spore counting, and sperm counting.