PBT Screw Caps for DURAN® Laboratory Glass Bottles, Wheaton, DWK Life Sciences
Catalog #: 82029-048
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:Screw Cap
- Cat. no.:82029-048
- Cap Color:Red
- Thread Size:GL 45
- Supplier No.:2924028
Specifications
About this item
The cap is available with a PTFE coated silicone seal or as a screw connection cap with aperture.
- Autoclavable
- Chemically resistant
- Temperature resistant to 180°C
Customers Who Bought This Also Bought