Nalgene® Biotainer® Polycarbonate Carboys, Thermo Scientific
Catalog #: 82028-472
Supplier: Thermo Fisher Scientific
Specifications
- Capacity:10 L (2.6 gal.)
- Description:Biotainer® Carboy without Handle
- Cat. no.:82028-472
- Supplier No.:3410-42
Specifications
About this item
Sterile carboys are molded from blue-tinted polycarbonate and provide safe storage for contents at temperatures from −85 °C (−121 °F) to near boiling
- Space-saving square shape
Carboys meet USP-VI and are noncytotoxic and nonpyrogenic. Gamma irradiated to a sterility assurance level of 10⁻⁶.
Ideal for storage and transport of media, sera, antigens, and other biological reagents. Carboys feature molded graduations in liters and gallons and indented, ribbed handgrips.
Ordering information: Each carboy is supplied with a 48mm screw cap with a silicone liner. Individually bagged.
Customers Who Bought This Also Bought