VWR® Safety Bulb Pipette Filler
Catalog #: 82024-574
Supplier: VWR International
Specifications
- Capacity:15 ml
- Cat. no.:82024-574
Specifications
About this item
Designed for use with small and large pipettes, bulb features a white plastic attachment with a silicone conical adapter for safe and secure attachment to pipettes.
- Easy application
- Secure attachment to pipettes
- Simple operation
- Suction action for drawing liquids
Attachment does not require twisting or rough pipette handling. Simply squeeze bulb and gently press pipette to form seal. Liquid is drawn by suction; lift away bulb to control fluid dispensing with index finger. Supplied with elastic band for hanging on bottles or wrist.
Customers Who Bought This Also Bought