VWR® Acrylic Pipettor Stands
Catalog #: 82024-540
Supplier: VWR International
Specifications
- Description:3-Place Stand
- Cat. no.:82024-540
Specifications
About this item
Convenient three-, four-, and six-place stands are designed to hold most major brands of pipettors.
- Clear acrylic construction
- Designed to hold most major brands of pipettors
They are constructed of clear acrylic and accommodate a variety of laboratory applications and needs. Upper slots are 25mm (1") wide and lower slots are 20mm (13/16") wide.
Customers Who Bought This Also Bought